Learn More
5alpha-Cholestane-2,2,3,3,4,4-d6, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 5 A-Cholestane-2,2,3,3,4,4-d6 |
| CAS | 358731-18-7 |
| Molecular Formula | C27H42D6 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C27H48/c1-19(2)9-8-10-20(3)23-14-15-24-22-13-12-21-11-6-7-17-26(21,4)25(22)16-18-27(23,24)5/h19-25 H,6-18H2,1-5H3/t20-,21-,22+,23-,24+,25+,26+,27-/m1/s1/i6D2,7D2,11D2 |
| SMILES | C[C@@]12 [C@]([C@]3([C@@]([C@]4(C)[C@@](CC3)(C(C(C(C4)([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])[H])(CC1)[H])[H])(CC[C@@]2([C@@H](CCCC(C)C)C)[H])[H] |
| IUPAC Name | (5 R,8 R,9 S,10 S,13 R,14 S,17 R)-10,13-dimethyl-17-((R)-6-methylheptan-2-yl)hexadecahydro-1 H-cyclopenta[a]phenanthrene-2,2,3,3,4,4-d6 |
| Molecular Weight (g/mol) | 378.72 |
| Formula Weight | 378.41 g/mol |
| Percent Purity | 95 atom % D, min 98% Chemical Purity |