Learn More
5alpha-Cholestan-3beta-ol, 98%
Description
5alpha-Cholestan-3beta-ol is used as a standard in lipid analysis using HPLC. It acts as a derivitized steroid compound. It is used in agrochemical, pharmaceutical and dyestuff field .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5alpha-Cholestan-3beta-ol is used as a standard in lipid analysis using HPLC. It acts as a derivitized steroid compound. It is used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
| Melting Point | 140°C to 144°C |
| Quantity | 25 g |
| Beilstein | 2418594 |
| Merck Index | 14,2200 |
| Solubility Information | Insoluble in water. |
| Optical Rotation | +24° (c=1 in Chloroform) |
| Formula Weight | 388.68 |
| Percent Purity | 98% |
| Chemical Name or Material | 5alpha-Cholestan-3beta-ol |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.