missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Androstan-17beta-ol-3-one-1,2,2,4,4,5-d6, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Androstanolone-1,2,2,4,4,5-D6 |
| Molecular Formula | C19 D6 H24 O2 |
| Quantity | 0.01 g |
| Synonym | 5α-Androstan-17β-ol-3-one-1,2,2,4,4,5-d6, (5α,17β)-17-Hydroxyandrostan-3-one-1,2,2,4,4,5-d6, Androstanolone-d6, Androstanolone-1,2,2,4,4,5-d6, Androstanolone-1,2,2,4,4,5-D6, 5α-Dihydrotestosterone-1,2,2,4,4,5-d6 |
| InChI Formula | InChI=1 S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12,14-17,21 H,3-11H2,1-2H3/t12-,14-,15-,16-,17-,18-,19-/m0/s1/i7D2,9 D,11D2,12 D/t9?,12-,14-,15-,16-,17-,18-,19- |
| SMILES | [2 H]C1C([2 H])([2 H])C(=O)C([2 H])([2 H])[C@]2([2 H])CC[C@H]3 [C@@H]4 CC[C@H](O)[C@@]4(C)CC[C@@H]3 [C@@]12 C |
| IUPAC Name | (5 S,8 R,9 S,10 S,13 S,14 S,17 S)-1,2,2,4,4,5-hexadeuterio-17-hydroxy-10,13-dimethyl-6,7,8,9,11,12,14,15,16,17-decahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 296.4773 |
| Formula Weight | 296.2622 g/mol |
| Percent Purity | 97 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction