missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Δ5a-11-Hydroxy-12-oxo Minocycline Disodium Salt, TRC
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | Delta5a-11-Hydroxy-12-oxo Minocycline Disodium Salt |
| Molecular Formula | C23 H23 N3 O7 . 2Na |
| Quantity | 5 mg |
| Synonym | Δ5a-11-Hydroxy-12-oxo Minocycline Disodium Salt, 4,7-Bis(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-1,12-dioxo-2-naphthacenecarboxamide Disodium Salt, 5a,6-Anhydro Minocycline Disodium Salt |
| InChI Formula | InChI=1S/C23H25N3O7.2Na/c1-25(2)12-5-6-13(27)15-10(12)7-9-8-11-17(26(3)4)19(29)16(22(24)32)21(31)23(11,33)20(30)14(9)18(15)28;;/h5-7,11,17,27-29,33H,8H2,1-4H3,(H2,24,32);;/q;2*+1/p-2/t11-,17-,23-;;/m0../s1 |
| SMILES | [Na+].[Na+].CN(C)[C@H]1[C@@H]2Cc3cc4c(ccc([O-])c4c(O)c3C(=O)[C@]2(O)C(=O)C(=C1[O-])C(=O)N)N(C)C |
| IUPAC Name | disodium;(6aS,7S,10aS)-9-carbamoyl-4,7-bis(dimethylamino)-10a,12-dihydroxy-10,11-dioxo-6a,7-dihydro-6H-tetracene-1,8-diolate |
| Molecular Weight (g/mol) | 499.42 |
| Formula Weight | 499.1331 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction