missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadyl Meso-Tetraphenylporphine, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Vanadyl Meso-Tetraphenylporphine |
| CAS | 14705-63-6 |
| Molecular Formula | C44 H28 N4 O V |
| Quantity | 50 mg |
| Synonym | Vanadium','oxo[5,10,15,20-tetraphenyl-21H,23H-porphinato(2-)-κN21,κN22,κN23,κN24]-','(SP-5-12)- (9CI','ACI), (SP-5-12)-Oxo[5,10,15,20-tetraphenyl-21H,23H-porphinato(2-)-κN21,κN22,κN23,κN24]vanadium (ACI), 21H,23H-Porphine','5,10,15,20-tetraphenyl-','vanadium complex (ZCI), 21H,23H-Porphine','vanadium deriv. (ZCI), Porphine','tetraphenyl-','V complex (6CI), Vanadium','oxo[5,10,15,20-tetraphenyl-21H,23H-porphinato(2-)-N21,N22,N23,N24]-','(SP-5-12)- (ZCI), Vanadium','oxo[5,10,15,20-tetraphenylporphinato(2-)]- (8CI), Vanadium','oxo[α,β,γ,Δ-tetraphenylporphinato(2-)]- (7CI), (5,10,15,20-Tetraphenyl-21H,23H-porphine)vanadium(IV) oxide, 5,10,15,20-tetraphenyl-21 H,23H-porphine vanadium(IV) oxide, Oxo(5,10,15,20-tetraphenylporphinato)vanadium, Oxo(tetraphenylporphyrinato)vanadium, Oxo[5,10,15,20-tetraphenylporphinato(2-)]vanadium, Oxovanadium(IV) tetraphenylporphyrin, Vanadyl 5,10,15,20-Tetraphenylporphyrin, Vanadyl tetraphenylporphine, Vanadyltetraphenylporphyrin |
| InChI Formula | InChI=1S/C44H28N4.O.V/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;;/h1-28H;;/q-2;;+2/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;; |
| SMILES | O=[V+2]123N4=C5C=CC4=C(C6=CC=C(C(=C7C=CC(=N17)C(=C8C=CC(=C5c9ccccc9)[N-]28)c%10ccccc%10)c%11ccccc%11)[N-]36)c%12ccccc%12 |
| Molecular Weight (g/mol) | 679.66 |
| Formula Weight | 679.1703 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction