missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine-d9 N-Oxide, TRC

Product Code. 30451016
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30451016

Brand: Toronto Research Chemicals TRCT79579250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylamine N-Oxide d9
CAS 1161070-49-0
Purity Grade Notes HPLC
Molecular Formula C3 D9 N O
Quantity 50 mg
Synonym Methan-d3-amine','N,N-di(methyl-d3)-','N-oxide (ACI), Trimethyl-d9-amine N-Oxide, Trimethylamine-d9 N-Oxide, Trimethylamine N-Oxide D9, N,N-Di(methyl-d3)methan-d3-amine N-oxide
InChI Formula InChI=1S/C3H9NO/c1-4(2,3)5/h1-3H3/i1D3,2D3,3D3
SMILES [2H]C([2H])([2H])[N+]([O-])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name 1,1,1-trideuterio-N,N-bis(trideuteriomethyl)methanamine oxide
Molecular Weight (g/mol) 84.16
Formula Weight 84.1249
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.