Learn More
Topotecan Hydrochloride, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Topotecan Hydrochloride |
| CAS | 119413-54-6 |
| Molecular Formula | C23 H23 N3 O5 . Cl H |
| Quantity | 50 mg |
| Synonym | (4S)-10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione Hydrochloride (1:1), 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione','10-[(dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-','monohydrochloride','(4S)- (9CI), 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione','10-[(dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-','monohydrochloride','(S)-, (S)-10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[l,2-b]quinoline-3,14(4H,12H)-dione monohydrochloride, Hycamtin, NSC 609669, Nogitecan hydrochloride, SKF 104864A, SKFS 104864A, Topotecan chlorohydrate, Topotecan hydrochloride, Xinze |
| InChI Formula | InChI=1S/C23H23N3O5.ClH/c1-4-23(30)16-8-18-20-12(9-26(18)21(28)15(16)11-31-22(23)29)7-13-14(10-25(2)3)19(27)6-5-17(13)24-20;/h5-8,27,30H,4,9-11H2,1-3H3;1H/t23-;/m0./s1 |
| SMILES | Cl.CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5c(CN(C)C)c(O)ccc5nc34)C2=O |
| IUPAC Name | (19S)-8-[(dimethylamino)methyl]-19-ethyl-7,19-dihydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.02,11.04,9.015,20]henicosa-1(21),2,4(9),5,7,10,15(20)-heptaene-14,18-dione;hydrochloride |
| Molecular Weight (g/mol) | 457.91 |
| Formula Weight | 457.1404 |