Learn More
Topotecan-d5, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Topotecan-d5 |
| CAS | 1133355-98-2 |
| Molecular Formula | C23H18D5N3O5 |
| Quantity | 50 mg |
| Synonym | (4S)-10-[(Dimethylamino)methyl]-4-(ethyl-d5)-4,9-dihydroxy-1H-pyrano[3’,4’,6-7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, NSC-609669-d5, SKF-104864A-d5, Hycamtin-d5 |
| InChI Formula | InChI=1S/C23H23N3O5/c1-4-23(30)16-8-18-20-12(9-26(18)21(28)15(16)11-31-22(23)29)7-13-14(10-25(2)3)19(27)6-5-17(13)24-20/h5-8,27,30H,4,9-11H2,1-3H3/t23-/m0/s1/i1D3,4D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5c(CN(C)C)c(O)ccc5nc34)C2=O |
| Molecular Weight (g/mol) | 426.48 |
| Formula Weight | 426.1952 |