Learn More
Tocris Bioscience™ OXA 06 dihydrochloride
GREENER_CHOICE
Product Code. 18720509
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18720509
Brand: Tocris Bioscience™ 5182/50
Potent ROCK inhibitor
OXA 06 dihydrochloride is a potent ROCK inhibitor (IC50 = 10 nM). Suppresses pMYPT1 and pCofilin levels in non-small cell lung carcinoma (NSCLC) cell lines. Also inhibits anchorage-independent growth of NSCLC cell lines in vitro.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1825455-91-1
Quantity 50 mg
Product Type OXA 06 dihydrochloride
Purity >98%
Molecular Weight (g/mol) 404.31
Inhibitors OXA 06 dihydrochloride