missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc (E)-Fluvoxamine, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | N-Fmoc (E)-Fluvoxamine |
| Molecular Formula | C30 H31 F3 N2 O4 |
| Quantity | 50 mg |
| Synonym | 7-[4-(Trifluoromethyl)phenyl]-5,12-dioxa-2,6-diazatridec-6-enoic Acid 9H-Fluoren-9-ylmethyl Ester |
| InChI Formula | InChI=1S/C30H31F3N2O4/c1-37-18-7-6-12-28(21-13-15-22(16-14-21)30(31,32)33)35-39-19-17-34-29(36)38-20-27-25-10-4-2-8-23(25)24-9-3-5-11-26(24)27/h2-5,8-11,13-16,27H,6-7,12,17-20H2,1H3,(H,34,36)/b35-28+ |
| SMILES | COCCCC\C(=N/OCCNC(=O)OCC1c2ccccc2c3ccccc13)\c4ccc(cc4)C(F)(F)F |
| IUPAC Name | 9H-fluoren-9-ylmethyl N-[2-[(E)-[5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxyethyl]carbamate |
| Molecular Weight (g/mol) | 540.573 |
| Formula Weight | 540.224 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction