missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fmoc-g-[Beta-(5-naphthyl Sulfonic Acid)-ethylenediamine]-L-glutamic Acid, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Fmoc-g-[beta-(5-naphthyl Sulfonic Acid)-ethylenediamine]-L-glutamic Acid |
| CAS | 193475-66-0 |
| Molecular Formula | C32 H31 N3 O8 S |
| Quantity | 50 mg |
| Synonym | Fmoc-G-[B-(5-Naphthyl Sulfonic Acid)-Ethylenediamine]-L-Glutamic Acid |
| InChI Formula | InChI=1S/C32H31N3O8S/c36-30(34-18-17-33-27-13-5-12-25-24(27)11-6-14-29(25)44(40,41)42)16-15-28(31(37)38)35-32(39)43-19-26-22-9-3-1-7-20(22)21-8-2-4-10-23(21)26/h1-14,26,28,33H,15-19H2,(H,34,36)(H,35,39)(H,37,38)(H,40,41,42)/t28-/m0/s1 |
| SMILES | OC(=O)[C@H](CCC(=O)NCCNc1cccc2c(cccc12)S(=O)(=O)O)NC(=O)OCC3c4ccccc4c5ccccc35 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-[2-[(5-sulfonaphthalen-1-yl)amino]ethylamino]pentanoic acid |
| Molecular Weight (g/mol) | 617.67 |
| Formula Weight | 617.1832 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction