Learn More
3'-Amino-2',3'-dideoxycytidine, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 3'-Amino-2',3'-dideoxycytidine |
| CAS | 84472-90-2 |
| Molecular Formula | C9H14N4O3 |
| Quantity | 50 mg |
| Synonym | 3'-Amino-2',3'-dideoxycytidine, NSC 365107 |
| InChI Formula | InChI=1S/C9H14N4O3/c10-5-3-8(16-6(5)4-14)13-2-1-7(11)12-9(13)15/h1-2,5-6,8,14H,3-4,10H2,(H2,11,12,15)/t5-,6+,8+/m0/s1 |
| SMILES | N[C@H]1C[C@@H](O[C@@H]1CO)N2C=CC(=NC2=O)N |
| IUPAC Name | 4-amino-1-[(2R,4S,5S)-4-amino-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
| Molecular Weight (g/mol) | 226.23 |