missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-N-Benzoyl-3-phenyl Isoserine Ethyl Ester, TRC

Product Code. 17925065
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17925065 50 mg 50mg
30137383 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17925065 Supplier Toronto Research Chemicals Supplier No. TRCB20855550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 153433-80-8
Molecular Formula C18 H19 N O4
Molecular Weight (g/mol) 313.35
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, ethyl ester, (αR,βS)-, Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, ethyl ester, [R-(R*,S*)]-, Ethyl (2R,3S)-3-(benzoylamino)-2-hydroxybenzenepropanoate, (2R,3S)-N-Benzoyl-3-phenylisoserine ethyl ester, Ethyl (2R,3S)-3-Benzamido-2-hydroxy-3-phenylpropanoate
IUPAC Name ethyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoate
SMILES CCOC(=O)[C@H](O)[C@@H](NC(=O)c1ccccc1)c2ccccc2

Specifikationer

Quantity 50 mg
InChI Formula InChI=1S/C18H19NO4/c1-2-23-18(22)16(20)15(13-9-5-3-6-10-13)19-17(21)14-11-7-4-8-12-14/h3-12,15-16,20H,2H2,1H3,(H,19,21)/t15-,16+/m0/s1
Formula Weight 313.1314
Percent Purity >95%
Chemical Name or Material N-Benzyloxymethylphenylisoserine ethylester
Grade HPLC
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.