missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Alloy, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Zinc, Alloy, 50g
Specifications
Specifications
| Chemical Name or Material | Tin/Cadmium/Lead/Chromium/Iron/Silicon/Nickel/Manganese/Magnesium/Copper/Aluminium |
| CAS | 7440-66-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Tin 0.0056 %/Cadmium 0.0058 %/Lead 0.0064 %/Chromium 0.009 %/Iron 0.022 %/Silicon 0.022 %/Nickel 0.0249 %/Manganese 0.044 %/Magnesium 0.093 %/Copper 1.122 %/Aluminium 4.35 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction