missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASRegistryNumber1t Iron, Pig Iron High, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
CASRegistryNumber1t Iron, Pig Iron High, 50g
Specifications
Specifications
| Chemical Name or Material | Zinc/Lead/Arsenic/Zirconium/Tungsten/Tin/Copper/Cobalt/Niobium/Molybdenum/Nickel/Phosphorus/Chromium/Vanadium/Manganese/Sulfur/Aluminium/Titanium/Silicon/Carbon/Iron |
| CAS | 7439-89-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Zinc -105 ppm/Lead -12 ppm/Arsenic 0.0015 %/Zirconium 0.0039 %/Tungsten 0.004 %/Tin 0.0075 %/Copper 0.0113 %/Cobalt 0.018 %/Niobium 0.019 %/Molybdenum 0.027 %/Nickel 0.04 %/Phosphorus 0.05 %/Chromium 0.088 %/Vanadium 0.108 %/Manganese 0.127 %/Sulfur 0.141 %/Aluminium 0.2 %/Titanium 0.42 %/Silicon 1.6 %/Carbon 4.42 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction