Learn More
Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits
Product Code. 15320762
Click to view available options
Diameter (Metric):
6.5 mm
16 mm
29 mm
67 mm
76 mm
Length (Metric):
0.32 mL/cm vol./length
2 mL/cm
34 mL/cm vol./length
46 mL/cm vol./length
5 m
6.4 mL/cm
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Each
Unit Size:
1m
5m
Each
Product Code. 15320762
Brand: Spectrum™ 132 680T1
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
  • 1m or 5m of Spectra/Por 2 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
  • For higher permeability applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)
Specifications
MWCO 12 to 14 kDa
Diameter (Metric) 29 mm
Length (Metric) 6.4 mL/cm
Quantity 1 m Tubing/Kit