Learn More
Chem-Lab 500ML Lead standard solution (Spectro ECON)(1.599 g Pb(NO3)2 / l 2% HNO3), for AAS
Brand: Chem-Lab CL01.1226.0500
500ML Lead standard solution (Spectro ECON)(1.599 g Pb(NO3)2 / l 2% HNO3), for AAS
Safety and Handling
- Lead standard solution 1000 µg/ml
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contain: Nitric acid, Lead(II) nitrate