Learn More
Ecgonidine Hydrochloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Anhydroecgonine Hydrochloride |
| CAS | 74242-55-0 |
| Molecular Formula | C9 H13 N O2 . Cl H |
| Quantity | 500 mg |
| Synonym | Anhydroecgonine Hydrochloride, (1R)-8-Methyl-8-azabicyclo[3.2.1]oct-2-ene-2-carboxylic acid hydrochloride, (1R,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-2-ene-2-carboxylic acid hydrochloride, (-)-Ecgonidine hydrochloride, Anhydroecgonine hydrochloride |
| InChI Formula | InChI=1S/C9H13NO2.ClH/c1-10-6-2-4-7(9(11)12)8(10)5-3-6;/h4,6,8H,2-3,5H2,1H3,(H,11,12);1H/t6-,8-;/m1./s1 |
| SMILES | Cl.CN1[C@H]2CC[C@@H]1C(=CC2)C(=O)O |
| IUPAC Name | (1S,5R)-8-methyl-8-azabicyclo[3.2.1]oct-3-ene-4-carboxylic acid;hydrochloride |
| Molecular Weight (g/mol) | 203.67 |
| Formula Weight | 203.0713 |