missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoxynil Octanoate, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Bromoxynil-octanoate |
| CAS | 1689-99-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H17 Br2 N O2 |
| Quantity | 500 mg |
| Synonym | Octanoic acid, ester with 3,5-dibromo-4-hydroxybenzonitrile (7CI,8CI), 2,6-Dibromo-4-cyanophenyl octanoate, 3,5-Dibromo-4-hydroxybenzonitrile octanoate ester, 3,5-Dibromo-4-n-octanoyloxybenzonitrile, 3,5-Dibromo-4-octanoyloxybenzonitrile, 4-Cyano-2,6-dibromophenyl octanoate, Brominal W, Bromotril, Bromoxynil n-octanoyl ester, Bromoxynil octanoate, Bromoxynil octanoic acid ester, Buctril, Buctril 33.4EC, Certrol B, NPH 1320 |
| InChI Formula | InChI=1S/C15H17Br2NO2/c1-2-3-4-5-6-7-14(19)20-15-12(16)8-11(10-18)9-13(15)17/h8-9H,2-7H2,1H3 |
| SMILES | CCCCCCCC(=O)Oc1c(Br)cc(cc1Br)C#N |
| IUPAC Name | (2,6-dibromo-4-cyanophenyl) octanoate |
| Molecular Weight (g/mol) | 403.11 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction