missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-chloroethyl)aminophosphoric Dichloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Bis(2-chloroethyl)aminophosphoric Dichloride |
| CAS | 127-88-8 |
| Molecular Formula | C4 H8 Cl4 N O P |
| Quantity | 500 mg |
| Synonym | Phosphoramidic dichloride, N,N-bis(2-chloroethyl)- (ACI), N,N-Bis(2-chloroethyl)phosphoramidic dichloride (ACI), Phosphoramidic dichloride, bis(2-chloroethyl)- (7CI, 8CI, 9CI), (N,N-Di-2-chloroethyl) phosphoramidic dichloride, 2-Chloro-N-(2-chloroethyl)-N-dichlorophosphorylethanamine, Bis(2-chloroethyl)aminophosphoric dichloride, Bis(2-chloroethyl)aminophosphoryl dichloride, Bis(2-chloroethyl)phosphoramidic dichloride, N,N-Bis(2-chloroethyl)phosphamide dichloride, NSC 64119 |
| InChI Formula | InChI=1S/C4H8Cl4NOP/c5-1-3-9(4-2-6)11(7,8)10/h1-4H2 |
| SMILES | ClCCN(CCCl)P(=O)(Cl)Cl |
| IUPAC Name | 2-chloro-N-(2-chloroethyl)-N-dichlorophosphorylethanamine |
| Molecular Weight (g/mol) | 258.9 |
| Formula Weight | 256.9098 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction