missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androstanolone Acetate, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Androstanolone Acetate |
| CAS | 1164-91-6 |
| Molecular Formula | C21H32O3 |
| Quantity | 500 mg |
| Synonym | 5alpha-Dihydrotestosterone Acetate, Androstanolone Acetate, Dihydrotestosterone Acetate, NSC 73107, Stanolone Acetate, 17-O-Acetyldihydrotestosterone, 17beta-Acetoxy-5alpha-androstan-3-one, 17beta-Hydroxy-5alpha-androstan-3-one Acetate, 3-Oxo-5alpha-androstan-17beta-yl Acetate, 5alpha-Androstan-17beta-ol-3-one Acetate |
| InChI Formula | InChI=1S/C21H32O3/c1-13(22)24-19-7-6-17-16-5-4-14-12-15(23)8-10-20(14,2)18(16)9-11-21(17,19)3/h14,16-19H,4-12H2,1-3H3/t14-,16-,17-,18-,19-,20-,21-/m0/s1 |
| SMILES | CC(=O)O[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(5S,8R,9S,10S,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl] acetate |
| Molecular Weight (g/mol) | 332.48 |
| Formula Weight | 332.2351 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction