Learn More
500 NTU Ratio Turbidity Standard, Reagecon
Shop All Reagecon Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
500 mL
missing translation for 'unitSize'
500mL
Description
- Turbidity Reference Standard, Turbidity Reference Material.
- Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
- Certified
- NIST traceable
- Non-toxic, non-carcinogenic
- Highly accurate
- Stable
- EPA approved
Specifications
Specifications
| For Use With (Equipment) | Ratio Instruments |
| Description | Liquid format in HDPE Bottle GL45 |
| Accuracy | Accuracy ± 1% NTU |
| Turbidity | 500 NTU ± 1% |
| Volume (Metric) | 500 mL |