Learn More
Thermo Scientific™ Oxoid™ Sabouraud Dextrose Agar
Click to view available options
Fill Weight:
500-504 g
Format:
500 mL sirop bottle
Unit Size:
Pack of 10
Description
The media are slightly acidic, which helps to inhibit bacteria while being favorable to fungi.
- Sabouraud Dextrose Agar 10 x 500 mL: contain mycological peptone and glucose to support growth of fungi. The media are slightly acidic which helps to inhibit bacteria whilst being favourable to fungi
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2 to 25°C |
| Format | 500 mL sirop bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 500-504 g |
| Packaging Type | 10 sirop bottles with screw cap closures in a box. |
| Quantity | 10 x 500 mL |