missing translation for 'onlineSavingsMsg'
Learn More

TdB Labs AB 500 kDa DEAE-dextran for Anion-exchange Chromatography

Product Code. 30216494
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30216494 100 g 100g
30216443 10 g 10g
30216428 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30216494 Supplier TdB Labs AB Supplier No. DD500_100g
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DEAE-dextran 500 kDa is a high MW cationic polymer used in protein binding, chromatography, and complexation. Also studied as a veterinary adjuvant, where its strong positive charge aids antigen delivery and immune activation.

  • Product Overview: DEAE-dextran is a cationic, water-soluble polysaccharide modified with diethylaminoethyl groups. Known for its strong positive charge and high biocompatibility, DEAE-dextran is ideal for researchers and bioprocess engineers working with protein purification, nucleic acid complexation, or surface functionalization. Available across a wide molecular weight range to match different project needs.
  • Key Features:
    • Strong Cationic Properties: DEAE-modification provides consistent positive charge for robust binding to negatively charged biomolecules like proteins, DNA, and RNA.
    • Flexible Molecular Weight Range: Offered from 4 kDa up to 2000 kDa, giving options for fine-tuning binding strength, diffusion, and separation in your application.
    • High Solubility: Fully water-soluble powder allows rapid and easy preparation for chromatography, formulation, or complexation protocols.
    • Batch-to-Batch Consistency: Tight control over molecular weight and functionalization ensures reliable results across experiments and production runs.
    • Stable Storage: Dry powder format provides long shelf life without special storage conditions, helping to minimize lab downtime.
  • Why Choose DEAE-dextran:
    • For Protein Purification: DEAE-dextran is a proven choice for anion-exchange chromatography workflows, helping isolate or fractionate proteins under mild, biocompatible conditions.
    • For Complexation and Binding Studies: Ideal for creating complexes with nucleic acids or proteins to study binding kinetics, surface adsorption, or biomolecular interactions.
    • For Transfection Research: Historically used in mammalian cell DNA transfection experiments, DEAE-dextran still finds niche use where simple, non-viral gene delivery is studied.
    • For Surface Modification: Researchers functionalizing surfaces (e.g., biosensors, biomaterials) benefit from DEAE-dextran's ability to introduce stable, positive surface charges.
    • For Vaccine Adjuvant Studies: Studied as a component in veterinary vaccine formulations to enhance immune system targeting by improving antigen presentation.
  • Typical Applications:
    • Ion-exchange chromatography for proteins and nucleic acids
    • Formation of electrostatic complexes for analytical and purification purposes
    • Historical and research-based DNA transfection in mammalian cells
    • Surface coating and modification in bioengineering projects
    • Supportive studies in vaccine and adjuvant formulation development
  • Technical Details:
    • Molecular Weights: 4 kDa, 20 kDa, 40 kDa, 70 kDa, 150 kDa, 500 kDa, and 2000 kDa available.
    • Charge Density: Approximately 0.1 to 0.2 DEAE groups per glucose unit (depending on the grade).
    • Appearance: White to off-white dry powder, highly soluble in water.
  • Storage and Handling:
    • Store dry at room temperature in a sealed container to maintain stability.
    • Protect from moisture to preserve full activity over long-term storage.
  • Availability:
    • Supplied as research-grade powder, suitable for laboratory use and pilot-scale process development.
    • Available in small packs for R and D or bulk volumes for production environments.

Specifications

Cell Type HEK293
Format Vial
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Species Human
Product Type Transfection Reagent
Quantity 100 g
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.