Få mere at vide
Få mere at vide
Labcon™ MetalFree™ 50 mL Polypropylene Centrifuge Tubes
Klik for at se tilgængelige muligheder
Packaging:
25 Tubes/Low Waste Fiber Rack, 20 Racks/Cs.
25 Tubes/Rack, 20 Rcks/Cs
50 Tubes/Bag, 10 Bags/Cs.
Pakningsstørrelse:
Pack of 500
Beskrivelse
- Conical centrifuge tube
- With attached flat screw caps
- Certified free of metals to 1 part per billion of 43 common metals and elements
- Caps are pure natural polyethylene
- Patented anti-smear writing surface
- ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Polypropylene USP Class VI resin
- Centrifuging to 17,000 x G
- Freezable to -80°C
- Autoclavable
- Capacity: 50 mL
- Natural colorless caps
- Made in USA
- Elements tested: Aluminium, Antimony, Arsenic, Barium,, Beryllium, Bismuth, Cadnium, Caesium, Calcium, Chromium, Cobalt, Copper, Gallium, Germanium, Gold, Iron, Lead , Lithium, Magnesium, Manganese, Mercury, Molebdenum, Nickel, Niobium, Phosphorus, Potassium, Rhenium, Rubidium, Selenium, Silicon, Silver, Sodium, Strontium, Tantalum, Thallium, Tin, Titanium, Tungsten, Vanadium, Yttrium, Zinc, Zirconium
Tekniske data
Tekniske data
| Certifications/Compliance | ISO 9001:2015, FDA CFR21 GMP |
| Closure Material | Polyethylene |
| Closure Color | Natural |
| Capacity (Metric) | 50 mL |
| Disposable | Single-use |
| Material | Polypropylene |
| For Use With | Centrifuges, rotators, shakers, ICP-MS |
| No. per Pack | 25/Pk. |
| Packaging | 25 Tubes/Rack, 20 Rcks/Cs |
| Sterility | Non-sterile |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion