Learn More
Tocris Bioscience™ RHPS 4 methosulfate
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
RHPS 4 methosulfate is a telomerase inhibitor. Inhibits growth of medulloblastoma and glioblastoma cells in vitro. Induces telomere injury and apoptosis, and reduces growth in CG5 breast cancer cell xenografts in mice. Sensitizes radiation resistant glioblastoma cell lines to radiation.
Specifications
Specifications
| Chemical Name or Material | 3,11-Difluoro-6,8,13-trimethylquino[4,3,2-kl]acridinium methylsulfate |
| CAS | 390362-78-4 |
| Quantity | 50 mg |
| Target | Telomerase Inhibitors |
| Molecular Formula | C22H17F2N2.CH3O4S |
| Formula Weight | 458.48 |
| Purity | >98% |