Learn More
5-Trifluoromethyl-1H-1,2,4-triazole-3-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Trifluoromethyl-1H-1,2,4-triazole-3-carboxylic Acid |
| CAS | 944897-41-0 |
| Molecular Formula | C4 H2 F3 N3 O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C4H2F3N3O2/c5-4(6,7)3-8-1(2(11)12)9-10-3/h(H,11,12)(H,8,9,10) |
| SMILES | OC(=O)c1nnc([nH]1)C(F)(F)F |
| IUPAC Name | 5-(trifluoromethyl)-4H-1,2,4-triazole-3-carboxylic acid |
| Molecular Weight (g/mol) | 181.07 |
| Formula Weight | 181.0099 |