Learn More
5-(tert-Butyl)isobenzofuran-1,3-dione, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 32703-79-0 |
| Molecular Formula | C12H12O3 |
| MDL Number | MFCD00060127 |
| Quantity | 1 g |
| SMILES | O=C1OC(C2=C1C=CC(C(C)(C)C)=C2)=O |
| Molecular Weight (g/mol) | 204.22 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-(tert-Butyl)isobenzofuran-1,3-dione |