Learn More
5-(t-Boc-amino)-1-pentyl-p-toluenesulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(t-Boc-amino)-1-pentyl-p-toluenesulfonate |
| CAS | 118811-34-0 |
| Molecular Formula | C17 H27 N O5 S |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C17H27NO5S/c1-14-8-10-15(11-9-14)24(20,21)22-13-7-5-6-12-18-16(19)23-17(2,3)4/h8-11H,5-7,12-13H2,1-4H3,(H,18,19) |
| SMILES | Cc1ccc(cc1)S(=O)(=O)OCCCCCNC(=O)OC(C)(C)C |
| IUPAC Name | 5-[(2-methylpropan-2-yl)oxycarbonylamino]pentyl 4-methylbenzenesulfonate |
| Molecular Weight (g/mol) | 357.46 |
| Formula Weight | 357.161 |