Learn More
5-Phenylhydantoin, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Phenylhydantoin |
| CAS | 89-24-7 |
| Molecular Formula | C9 H8 N2 O2 |
| Quantity | 500 mg |
| Synonym | 2,4-Imidazolidinedione','5-phenyl-, Hydantoin','5-phenyl- (6CI,7CI,8CI), 5-Phenyl-2,4-imidazolidinedione, (±)-5-Phenylhydantoin, 5-Phenylhydantoin, 5-Phenylimidazoline-2,4-dione, DL-5-Phenylhydantoin, NSC 27302, NSC 40885, NSC 51847, Phenylhydantoin |
| InChI Formula | InChI=1S/C9H8N2O2/c12-8-7(10-9(13)11-8)6-4-2-1-3-5-6/h1-5,7H,(H2,10,11,12,13) |
| SMILES | O=C1NC(C(=O)N1)c2ccccc2 |
| IUPAC Name | 5-phenylimidazolidine-2,4-dione |
| Molecular Weight (g/mol) | 176.17 |
| Formula Weight | 176.0586 |