Learn More
5 NTU Ratio Turbidity Standard, Reagecon
BIOPHARMA
Product Code. 11566573 Shop All Reagecon Products
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11566573 500 mL 500mL
11778455 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11566573 Supplier Reagecon Supplier No. CRSR5500
  • ONLINE EXCLUSIVE PRICE
    167.05€
    151.62€ / 500mL
    Save 15.43€
    9% Off
 Restricted Item
In Stock
A suspension of polymer microspheres designed to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards
  • Guaranteed stability throughout shelf life even after opening bottle
  • ± 1% accuracy
  • NIST traceable
  • US EPA approved
Specifications
Type Turbidity Standard
For Use With (Equipment) Ratio Instruments
Form Liquid
Accuracy ± 1% NTU
Volume (Metric) 500 mL
Packaging Type HDPE Bottle GL45