Learn More
5-Norbornene-2-exo,3-exo-dimethanol, 97%
Product Code. 15420218
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420218 1 g 1g
15430218 5 g 5g
15440218 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15420218 Supplier Thermo Scientific Alfa Aesar Supplier No. H53422.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
CAS: 699-95-6 | C9H14O2 | 154.209 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 699-95-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00799568
InChI Key IGHHPVIMEQGKNE-ROVKLQMOSA-N
Synonym bicyclo 2.2.1 hept-5-ene-2-exo,3-exo-dimethanol
PubChem CID 73995587
IUPAC Name [(1R,3S,4S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol
SMILES C1C2C=CC1C(C2CO)CO
Specifications
Density 1.027
Boiling Point 97°C (20 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.523
Quantity 1 g
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 5-Norbornene-2-exo,3-exo-dimethanol