Learn More
5-(N-tert-Butoxycarbonylamino)salicylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(N-tert-Butoxycarbonylamino)salicylic Acid |
| CAS | 135321-95-8 |
| Molecular Formula | C12 H15 N O5 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C12H15NO5/c1-12(2,3)18-11(17)13-7-4-5-9(14)8(6-7)10(15)16/h4-6,14H,1-3H3,(H,13,17)(H,15,16) |
| SMILES | CC(C)(C)OC(=O)Nc1ccc(O)c(c1)C(=O)O |
| IUPAC Name | 2-hydroxy-5-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid |
| Molecular Weight (g/mol) | 253.25 |
| Formula Weight | 253.095 |