missing translation for 'onlineSavingsMsg'
Learn More

5-(morpholinomethyl)isoxazole-3-carbaldehyde, Thermo Scientific™

Product Code. p-4391433
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10179909 250 mg Amber Glass Bottle 250mg
10637580 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10179909 Supplier Thermo Scientific Maybridge Supplier No. CC70804CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 893749-85-4
Molecular Formula C9H12N2O3
Molecular Weight (g/mol) 196.206
InChI Key AGLGFPIEVWZSLU-UHFFFAOYSA-N
Synonym 5-morpholinomethyl isoxazole-3-carbaldehyde, 5-morpholin-4-ylmethyl-1,2-oxazole-3-carbaldehyde, 5-morpholin-4-ylmethyl isoxazole-3-carbaldehyde, 5-morpholin-4-yl methyl-1,2-oxazole-3-carbaldehyde
PubChem CID 20099059
IUPAC Name 5-(morpholin-4-ylmethyl)-1,2-oxazole-3-carbaldehyde
SMILES C1COCCN1CC2=CC(=NO2)C=O

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 196.21
Percent Purity 97%
Chemical Name or Material 5-(morpholinomethyl)isoxazole-3-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.