Learn More
5-Methylthiophene-3-carbaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Methylthiophene-3-carbaldehyde |
| CAS | 29421-72-5 |
| Molecular Formula | C6 H6 OS |
| Quantity | 50 mg |
| Synonym | 4-Methyl-(9CI)-1H-imidazole-2-carboxylic Acid, 4-Methyl-1H-imidazole-2-carboxylic Acid, 4-Methylimidazole-2-carboxylic Acid |
| InChI Formula | InChI=1S/C6H6OS/c1-5-2-6(3-7)4-8-5/h2-4H,1H3 |
| SMILES | Cc1cc(C=O)cs1 |
| IUPAC Name | 5-methylthiophene-3-carbaldehyde |
| Molecular Weight (g/mol) | 126.176 |
| Formula Weight | 126.0139 |