Learn More
5-Methylpyrimidine, 98%, Thermo Scientific™
Product Code. 15254844 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15254844 250 mg Glass bottle 250mg
15264844 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15254844 Supplier Thermo Scientific Acros Supplier No. 450732500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 2036-41-1
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00047386
InChI Key TWGNOYAGHYUFFR-UHFFFAOYSA-N
Synonym pyrimidine, 5-methyl, 5-methyl-pyrimidine, pyrimidine, 5-methyl-6ci,7ci,8ci,9ci, 5-methyl-pyrimidin, pubchem7207, rarechem am la 0031, 5-methyl-1,3-diazine, acmc-1cnd2, 5-methylpyrimidine, 5-methylpyrimidine, 97+%
PubChem CID 74859
IUPAC Name 5-methylpyrimidine
SMILES CC1=CN=CN=C1
Specifications
Melting Point 33°C to 37°C
Boiling Point 152°C to 154°C
Flash Point 52°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble
Formula Weight 94.11
Percent Purity 98%
Chemical Name or Material 5-Methylpyrimidine
compliance-icons compliance-icons
Product Identifier
  • 5-Methylpyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Title
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.