Learn More
5-Methyl-2-methylamino-1,3-thiazole, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Methyl-2-methylamino-1,3-thiazole |
| CAS | 52829-91-1 |
| Molecular Formula | C5H8N2S |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C5H8N2S/c1-4-3-7-5(6-2)8-4/h3H,1-2H3,(H,6,7) |
| SMILES | CNc1ncc(C)s1 |
| IUPAC Name | N,5-dimethyl-1,3-thiazol-2-amine |
| Molecular Weight (g/mol) | 128.2 |
| Formula Weight | 128.0408 |