Learn More
5-Methoxytryptamine-alpha,alpha,beta,beta-d4, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 5-Methoxytryptamine-alpha,alpha,beta,beta-d4 |
| CAS | 96236-05-4 |
| Molecular Formula | C11 D4 H10 N2 O |
| Quantity | 0.05 g |
| Synonym | 1 H-Indole-3-ethan-α,α,β,β-d4-amine, 5-methoxy- (9 CI), 5-Methoxytryptamine-α,α,β,β-D4, 5-Methoxytryptamine-α,α,β,β-d4, 5-Methoxytryptamine-d4, 5-Methoxy-1 H-indole-3-ethan-α,α,β,β-d4-amine |
| InChI Formula | InChI=1 S/C11H14N2O/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11/h2-3,6-7,13 H,4-5,12H2,1H3/i4D2,5D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])c1c[nH]c2ccc(OC)cc12 |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-(5-methoxy-1 H-indol-3-yl)ethanamine |
| Molecular Weight (g/mol) | 194.266 |
| Formula Weight | 194.136 g/mol |
| Show More |