missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxypiperonal, 97%

Product Code. 11378035
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11378035

Brand: Thermo Scientific Alfa Aesar L08213.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5780-07-4 | C9H8O4 | 180.16 g/mol

5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with air, oxidizing agents. Store under dry inert gas. Store away from air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5780-07-4
MDL Number MFCD00016900

Specifications

Melting Point 126°C to 132°C
Molecular Formula C9H8O4
Quantity 5 g
Beilstein 384077
Sensitivity Air Sensitive
Synonym myristicin aldehyde, 3-methoxy-4,5-methylenedioxybenzaldehyde, 3-methoxypiperonal, 1,3-benzodioxole-5-carboxaldehyde, 7-methoxy, 5-methoxypiperonal, 7-methoxy-benzo 1,3 dioxole-5-carbaldehyde, 7-methoxybenzo d 1,3 dioxole-5-carbaldehyde, 7-methoxy-2h-1,3-benzodioxole-5-carbaldehyde, 7-methoxybenzo-1,3-dioxole-5-carboxaldehyde, 7-methoxybenzo 1,3 dioxole-5-carbaldehyde
Solubility Information Insoluble in water.
InChI Key LOFRBHZYZCIOJO-UHFFFAOYSA-N
SMILES COC1=CC(=CC2=C1OCO2)C=O
IUPAC Name 7-methoxy-1,3-benzodioxole-5-carbaldehyde
Molecular Weight (g/mol) 180.16
PubChem CID 22016
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 5-Methoxypiperonal
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.