Learn More
5-Methoxycarbonyl Methyl Uridine, TRC
Product Code. 30481981
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481981 50 mg 50mg
30467828 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30481981 Supplier Toronto Research Chemicals Supplier No. TRCM26161550MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29428-50-0
Molecular Formula C12H16N2O8
Molecular Weight (g/mol) 316.26
Synonym 1,2,3,4-Tetrahydro-2,4-dioxo-1-β-D-ribofuranosyl-5-pyrimidineacetic Acid Methyl Ester, 5-Carboxymethyluridine Methyl Ester
SMILES O[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N(C=C2CC(OC)=O)C(NC2=O)=O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C12H16N2O8/c1-21-7(16)2-5-3-14(12(20)13-10(5)19)11-9(18)8(17)6(4-15)22-11/h3,6,8-9,11,15,17-18H,2,4H2,1H3,(H,13,19,20)/t6-,8-,9-,11-/m1/s1
Chemical Name or Material 5-Methoxycarbonyl Methyl Uridine