Learn More
5-Methoxy-2-formylphenylboronic acid, TRC
Product Code. 30476388
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476388 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30476388 Supplier Toronto Research Chemicals Supplier No. TRCM26342850MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methoxy-2-formylphenylboronic acid
CAS 40138-18-9
Molecular Formula C8H9BO4
Quantity 50 mg
Synonym B-(2-Formyl-5-methoxyphenyl)boronic Acid, 2-Formyl-5-methoxyphenylboronic Acid, 5-Methoxy-2-formylphenylboronic Acid
InChI Formula InChI=1S/C8H9BO4/c1-13-7-3-2-6(5-10)8(4-7)9(11)12/h2-5,11-12H,1H3
SMILES COc1ccc(C=O)c(c1)B(O)O
IUPAC Name (2-formyl-5-methoxyphenyl)boronic acid
Molecular Weight (g/mol) 179.97
Formula Weight 180.0594