Learn More
5-Hydroxypentanoic Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxypentanoic Acid Sodium Salt |
| CAS | 37435-69-1 |
| Molecular Formula | C5H9NaO3 |
| Quantity | 5 g |
| Synonym | Sodium 5-Hydroxypentanoate, Sodium 5-Hydroxyvalerate |
| InChI Formula | InChI=1S/C5H10O3.Na/c6-4-2-1-3-5(7)8;/h6H,1-4H2,(H,7,8);/q;+1/p-1 |
| SMILES | OCCCCC(O[Na])=O |
| IUPAC Name | sodium;5-hydroxypentanoate |
| Molecular Weight (g/mol) | 140.11 |
| Formula Weight | 140.0449 |