missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy Omeprazole Sodium Salt, TRC

Product Code. 30005094
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005094 2.5 mg 2.5mg
30010327 1 mg 1mg
30006484 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005094 Supplier Toronto Research Chemicals Supplier No. TRCH9488632.5MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215629-45-0
Molecular Formula C17H18N3NaO4S
Molecular Weight (g/mol) 383.4
SMILES [Na+].COc1ccc2[n-]c(nc2c1)S(=O)Cc3ncc(CO)c(OC)c3C

Specifications

Quantity 2.5 mg
Percent Purity >95%
Chemical Name or Material 5-Hydroxy Omeprazole Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.