Learn More
5-Hydroxy-3,4-dimethoxybenzoic acid, 97+%
Product Code. 11327565
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 11327565
Brand: Thermo Scientific Alfa Aesar L05708.MD
Request Bulk or Custom Format
CAS: 8-1-1916 | C9H10O5 | 198.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8-1-1916
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.17
MDL Number MFCD00002508
InChI Key WFIBQVFJXGQICQ-UHFFFAOYSA-N
Synonym 3,4-dimethoxy-5-hydroxybenzoic acid, 5-hydroxy-3,4-dimethoxybenzoic acid, 5-hydroxyveratric acid, 3,4-o-dimethylgallic acid, 5-hydroxy veratric acid, chembl85234, 3,4-dimethoxy-5-hydroxy benzoic acid, acmc-20an2p, 3-hydroxy-4 5-dimethoxybenzoic acid, 3-hydroxy-4,5-dimethoxy-benzoic acid
PubChem CID 74709
IUPAC Name 3-hydroxy-4,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(=C1OC)O)C(=O)O
Specifications
Melting Point 194°C to 197°C
Quantity 250 mg
Beilstein 2115310
Formula Weight 198.17
Percent Purity ≥97%
Chemical Name or Material 5-Hydroxy-3,4-dimethoxybenzoic acid