Learn More
5-Hydroxy-3,4-dimethoxybenzaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxy-3,4-dimethoxybenzaldehyde |
| CAS | 29865-90-5 |
| Molecular Formula | C9H10O4 |
| Quantity | 2.5 g |
| Synonym | 5-Hydroxy-veratraldehyde, 3,4-Dimethoxy-5-hydroxybenzaldehyde, 3-Hydroxy-4,5-dimethoxybenzaldehyde, 5-Hydroxy-3,4-dimethoxybenzaldehyde, 5-Hydroxyveratraldehyde |
| InChI Formula | InChI=1S/C9H10O4/c1-12-8-4-6(5-10)3-7(11)9(8)13-2/h3-5,11H,1-2H3 |
| SMILES | COc1cc(C=O)cc(O)c1OC |
| IUPAC Name | 3-hydroxy-4,5-dimethoxybenzaldehyde |
| Molecular Weight (g/mol) | 182.17 |
| Formula Weight | 182.0579 |