Learn More
5-Hydroxy-1-methylpyrazole, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxy-1-methylpyrazole |
| CAS | 33641-15-5 |
| Molecular Formula | C4H6N2O |
| Quantity | 1 g |
| Synonym | 1-Methyl-1H-pyrazol-5-ol, 1-Methylpyrazol-5-ol, 1-Methyl-5-hydroxy-1H-pyrazole, 1-Methyl-5-hydroxypyrazole, 5-Hydroxy-1-methyl-5-pyrazole |
| InChI Formula | InChI=1S/C4H6N2O/c1-6-4(7)2-3-5-6/h2-3,7H,1H3 |
| SMILES | Cn1nccc1O |
| IUPAC Name | 2-methylpyrazol-3-ol |
| Molecular Weight (g/mol) | 98.1 |
| Formula Weight | 98.048 |