Learn More
5-Hydroxy-1,4-naphthalenedione, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 481-39-0 |
| Molecular Formula | C10H6O3 |
| MDL Number | MFCD00001684 |
| Quantity | 25 g |
| SMILES | O=C(C1=C2C=CC=C1O)C=CC2=O |
| Molecular Weight (g/mol) | 174.15 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Hydroxy-1,4-naphthalenedione |