Learn More
5-Hydrazino-2(1H)-quinolinone Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Hydrazino-2(1H)-quinolinone Hydrochloride |
| CAS | 372078-45-0 |
| Molecular Formula | C9H10ClN3O |
| Quantity | 250 mg |
| Synonym | 5-Hydrazinyl-2(1H)-quinolinone Hydrochloride |
| InChI Formula | InChI=1S/C9H9N3O.ClH/c10-12-8-3-1-2-7-6(8)4-5-9(13)11-7;/h1-5,12H,10H2,(H,11,13);1H |
| SMILES | Cl.NNc1cccc2NC(=O)C=Cc12 |
| IUPAC Name | 5-hydrazinyl-1H-quinolin-2-one;hydrochloride |
| Molecular Weight (g/mol) | 211.65 |
| Formula Weight | 211.0512 |