Learn More
5-Fluoro-6-hydroxyhydro Uracil(Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | 5-Fluoro-6-hydroxyhydro Uracil(Mixture of Diastereomers) |
| CAS | 37103-91-6 |
| Molecular Formula | C4H5FN2O3 |
| Quantity | 50 mg |
| Synonym | 5-Fluorodihydro-6-hydroxy-2,4(1H,3H)-pyrimidinedione, NSC 528189 |
| InChI Formula | InChI=1S/C4H5FN2O3.H2O/c5-1-2(8)6-4(10)7-3(1)9;/h1-2,8H,(H2,6,7,9,10);1H2 |
| SMILES | O.OC1NC(=O)NC(=O)C1F |
| IUPAC Name | 5-fluoro-6-hydroxy-1,3-diazinane-2,4-dione;hydrate |
| Molecular Weight (g/mol) | 148.09 |
| Formula Weight | 166.039 |