Learn More
5-Fluoro-4-hydroxy-2-methoxypyrimidine, TRC
Product Code. 30407633
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407633 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30407633 Supplier Toronto Research Chemicals Supplier No. TRCF5917751G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Fluoro-4-hydroxy-2-methoxypyrimidine
CAS 1480-96-2
Molecular Formula C5 H5 F N2 O2
Quantity 1 g
Synonym Capecitabine Impurity 35
InChI Formula InChI=1S/C5H5FN2O2/c1-10-5-7-2-3(6)4(9)8-5/h2H,1H3,(H,7,8,9)
SMILES COC1=NC=C(F)C(=O)N1
IUPAC Name 5-fluoro-2-methoxy-1H-pyrimidin-6-one
Molecular Weight (g/mol) 144.1
Formula Weight 144.0335